Human Tagged ORF Clone Lentiviral Particle, PD-L1 (CD274)
Lieferant: ORIGENE TECHNOLOGIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
RC213071L3V
ORIGRC213071L3VEA 1180 EUR InStock
ORIGRC213071L3V
Human Tagged ORF Clone Lentiviral Particle, PD-L1 (CD274)
Vektoren, Plasmide und Bibliotheken
Lenti ORF particles, CD274 (Myc-DDK tagged) - Human CD274 molecule (CD274), 200 µl, >10⁷ TU/ml.
The molecular sequence of this clone aligns with the gene accession number as a point of reference only. However, individual transcript sequences of the same gene can differ through naturally occurring variations (e.g. polymorphisms), each with its own valid existence. This clone is substantially in agreement with the reference, but a complete review of all prevailing variants is recommended prior to use.
This clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge