Test kits and tablets for AQUAfast II colorimeters
Lieferant: THERMO ORION
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AC2001 AC2002 AC2009 AC2012 AC2016 AC2017 AC2027 AC2029 AC2030 AC2046 AC2048 AC2055 AC2060 AC2061 AC2065 AC2070 AC2071 AC2072 AC2078 AC2082 AC2096 AC2099 AC2098
ORIOAC2001EA 39.2 EUR InStock
ORIOAC2001 ORIOAC2002 ORIOAC2009 ORIOAC2012 ORIOAC2016 ORIOAC2017 ORIOAC2027 ORIOAC2029 ORIOAC2030 ORIOAC2046 ORIOAC2048 ORIOAC2055 ORIOAC2060 ORIOAC2061 ORIOAC2065 ORIOAC2070 ORIOAC2071 705-0750 ORIOAC2078 ORIOAC2082 ORIOAC2096 ORIOAC2099 705-0733
Test kits and tablets for AQUAfast II colorimeters
Umwelt-Prüfkits
Orion™ AQUAfast tablet reagents perform colorimetric measurements of common water parameters with convenient, rapidly dissolving tablets.
  • For use with ORION™ AQUAfast AQ4000, ORION™ AQUAfast AQ3070, ORION™ AQUAfast AQ3700, and discontinued ORION™ AQ2 series meters and other colorimeters
  • 50 to 100 tests per kit
Use Orion™ AQUAfast Tablet Reagents with Orion™ AQUAfast colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging. They are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide, and zinc.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge