Culture media in bags, ReadyBags™
Lieferant: THERMO FISHER DIAGNOSTICS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BM0104T BM0204T BM1104T BM9901A
OXOIBM0104TEA 225 EUR InStock
OXOIBM0104T OXOIBM0204T OXOIBM1104T OXOIBM9901A
Culture media in bags, ReadyBags™
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
ReadyBags™ are designed for use in food testing by laboratories with large numbers of samples requiring large volumes of broth media or diluent. Each ReadyBag™ contains 3 liters and is intended for use with an automatic dispenser. The ReadyBag™ is connected to tubing for standard laboratory equipment, such as gravimetric diluters or peristaltic pumps, by means of a connector. Connectors are available in two formats: autoclavable, re-usable stainless steel connectors or sterile, plastic, single-use connectors.
  • For large numbers of samples
  • Available in 3 L
  • Once empty, the bag can be disposed as normal laboratory waste
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge