TD0900A
TD0920A
TD0930A
TD0940A
TD0950A
TD0960A
OXOITD0900AEA
842
EUR
InStock
OXOITD0900A
OXOITD0920A
OXOITD0930A
OXOITD0940A
OXOITD0950A
OXOITD0960A
Latex toxin detection kits, RPLA
Mikrobiologische Testsysteme
Antigen-Antikörper-Tests
Ag-AK, Latex
The technique of reversed passive latex agglutination (RPLA) enables soluble antigen such as bacterial toxins to be detected in an agglutination assay. In a standard agglutination assay, soluble antibody reacts with particulate antigen such as bacterial cells. However, in a reversed agglutination assay the antibody, which is attached to particles, reacts with the soluble antigen. The particles (in this case, latex) do not themselves play a part in the reaction and they are therefore passive.
- The cross-linking of the latex particles by the specific antigen/antibody reaction results in the visible latex agglutination reaction