Latex toxin detection kits, RPLA
Lieferant: THERMO FISHER DIAGNOSTICS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TD0900A TD0920A TD0930A TD0940A TD0950A TD0960A
OXOITD0900AEA 842 EUR InStock
OXOITD0900A OXOITD0920A OXOITD0930A OXOITD0940A OXOITD0950A OXOITD0960A
Latex toxin detection kits, RPLA
Mikrobiologische Testsysteme Antigen-Antikörper-Tests Ag-AK, Latex
The technique of reversed passive latex agglutination (RPLA) enables soluble antigen such as bacterial toxins to be detected in an agglutination assay. In a standard agglutination assay, soluble antibody reacts with particulate antigen such as bacterial cells. However, in a reversed agglutination assay the antibody, which is attached to particles, reacts with the soluble antigen. The particles (in this case, latex) do not themselves play a part in the reaction and they are therefore passive.
  • The cross-linking of the latex particles by the specific antigen/antibody reaction results in the visible latex agglutination reaction
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge