Reagents for the determination of COD for photometers
Lieferant: PALINTEST
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PL456 PL454 PL450 PL460 PL462 PL464 PL452
PALIPL456EA 148 EUR InStock
PALIPL456 PALIPL454 PALIPL450 PALIPL460 PALIPL462 PALIPL464 PALIPL452
Reagents for the determination of COD for photometers
Umwelt-Prüfkits
Pre-dispensed liquid reagents for minimal handing with barcoded reagent labelling for ease of use in complete wastewater analysis.
  • Greater sensitivity in the lower part of the measurement range
  • Can be used with multiparameter photometer and heater block
Lieferumfang: Supplied in 16 mm tubes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge