PL456
PL454
PL450
PL460
PL462
PL464
PL452
PALIPL456EA
148
EUR
InStock
PALIPL456
PALIPL454
PALIPL450
PALIPL460
PALIPL462
PALIPL464
PALIPL452
Reagents for the determination of COD for photometers
Umwelt-Prüfkits
Pre-dispensed liquid reagents for minimal handing with barcoded reagent labelling for ease of use in complete wastewater analysis.
- Greater sensitivity in the lower part of the measurement range
- Can be used with multiparameter photometer and heater block
Lieferumfang: Supplied in 16 mm tubes.