Vorfilterkapsel, Mini Profile®
Lieferant: Pall Laboratory
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
12070 12071 12072 12073 12074
PALL12070EA 306 EUR InStock
PALL12070 PALL12071 PALL12072 PALL12073 PALL12074
Vorfilterkapsel, Mini Profile®
Filter Membranfilterkapseln
These are scaleable depth capsules for prefiltration and clarification. They are designed for small-volume production and scale-up evaluation.
  • High capacity depth medium is absolute particle rated for reliability
  • Broad chemical compatibility
  • Optimised for viscous fluids
  • Excellent for gel removal
Mini Profile II filters have a thick depth structure, providing high capacity for larger solids and gels as well as for fine particles.
Profile Star filters feature high area star-shaped pleated construction. The patented design combines the advantages of thick depth filters with those of traditional high area pleated filters.
PP filter media, core/end caps, and housing.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge