Anti-Porin (Ab-5) (185-197) Rabbit Polyclonal Antibody
Lieferant: Merck Millipore (Oncogene)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PC548-50
PC548-50EA 485 EUR InStock
PC548-50
Anti-Porin (Ab-5) (185-197) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Porin (Ab-5) (185-197)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge