Screw top waste and wash vial for autosampler
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
09923031
PERC09923031EA 4.3 EUR InStock
PERC09923031
Screw top waste and wash vial for autosampler
Vials
This screw top waste and wash vial is manufactured from type I borosilicate glass.
Zertifizierungen: Meets USP, JP, and EP Pharmacopeia requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge