PHR1158-1G
PHR1158-1GEA
87.7
EUR
InStock
PHR1158-1G
Pharmaceutical secondary standard, Benzocaine, certified reference material, Supelco®
Standards
Organische Standards
Traceable to USP 1054000.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Zertifizierungen: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.