Drucken
1-Step™, Pierce™ TMB-Blotting substrate solution
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
34018
PIER34018EA 191 EUR
PIER34018
1-Step™, Pierce™ TMB-Blotting substrate solution
Elektrophorese-Reagenzien Western Blotting Reagents
The 1-Step TMB-Blotting is a one component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry. TMB is highly sensitive and generates a blue-purple precipitate. This substrate is provided ready to use for maximum convenience.