Cleanroom mops, MicroMopp Plus
Lieferant: PFENNIG REINIGUNGSTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2700530
PPSA2700530EA 0 EUR InStock
PPSA2700530
Cleanroom mops, MicroMopp Plus
Mopps
Mops for wet and moist cleaning in single-stage technique.
  • For all smooth and hard floor coverings
  • Made up of high-tech-microfiber 100% PES
  • With backfilling, cover sheet, loop with TEC-polyester and pocket of harness for extremely tear-proof
For the use in ISO class 9.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge