Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SERPINA3 Mouse Monoclonal Antibody [clone: AACT/1452]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-604
PRSI33-604EA 773 EUR InStock
PRSI33-604
Anti-SERPINA3 Mouse Monoclonal Antibody [clone: AACT/1452]
Antikörper
Alpha 1-Antichymotrypsin, also called AACT and SERPINA3, inhibits the activity of certain proteases, such as cathepsin G that is found in neutrophils, and chymases found in mast cells, by cleaving them into a different shape or conformation. This activity protects some tissues, such as the lower respiratory tract, from damage caused by proteolytic enzymes. This protein is produced in the liver, and is an acute phase protein that is induced during inflammation. [Wiki].

Western blot: 0,5 - 1 µg/ml IHC (Paraffin): 0,5 - 1 µg/ml IF: 0,5 - 1 µg/ml Flow Cytometry: 0,5 - 1 µg/10e6 cells in 0,1 ml The stated application concentrations are sµggested starting amounts. Titration of the Alpha-1-Antichymotrypsin antibody may be required due to differences in protocols and secondary/substrate sensitivity.

Type: Primary
Antigen: SERPINA3
Clonality: Monoclonal
Clone: AACT/1452
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge