Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Cd40 Hamster Monoclonal Antibody [clone: HM40-3]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-026
PRSI76-026EA 628 EUR InStock
PRSI76-026
Anti-Cd40 Hamster Monoclonal Antibody [clone: HM40-3]
Antikörper
The HM40-3 monoclonal antibody reacts with mouse CD40, a 48 kDa type I glycoprotein known as Bp50. CD40 is part of the TNFR family and expressed on a subset of T cells, B cells, dendritic cells, and macrophages. It has been shown to be involved in the protection of B cells from apoptosis, B cell differentiation, co-stimulation, and isotype class-switching. The HM40-3 antibody blocks the binding of CD40L (CD154) and stimulates B and dendritic cell proliferation.

Type: Primary
Antigen: Cd40
Clonality: Monoclonal
Clone: HM40-3
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgM kappa
Reactivity: Mouse;Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge