Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ITGB3 Mouse Monoclonal Antibody [clone: VI-PL2]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-175
PRSI76-175EA 377 EUR InStock
PRSI76-175
Anti-ITGB3 Mouse Monoclonal Antibody [clone: VI-PL2]
Antikörper
The VI-PL2 monoclonal antibody reacts with CD61, a type I integral transmembrane glycoprotein also known as integrin beta 3. CD61 forms a complex with CD41 (platelet gpIIb) and CD51 (integrin alphaV). It is expressed on osteoclasts, fibroblasts, platelets, macrophages, and megakaryocytes and appears to bind to fibronectin, vitronectin, thrombospondin, and fibrinogen and vWF. The VI-PL2 antibody is reported to be cross-reactive with baboon, rhesus, and cynomolgus.

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: VI-PL2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge