Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Fcgr3; Fcgr2b Rat Monoclonal Antibody [clone: 2.4G2]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-576
PRSI76-576EA 884 EUR InStock
PRSI76-576
Anti-Fcgr3; Fcgr2b Rat Monoclonal Antibody [clone: 2.4G2]
Antikörper
The 2.4G2 monoclonal antibody specifically reacts with an epitope on the extracellular domain of the mouse CD16 (Fc gamma III) and CD 32 (Fc gamma II). CD16 and CD32 are low affinity receptors for the IgG Fc domain and are expressed by B lymphocytes, NK cells, kupffer cells, mast cells, monocytes, macrophages, granulocytes, immature thymocytes, neutrophils, and some activated mature T cells.The 2.4G2 antibody blocks the binding of immunoglobulins to CD16 and CD32, and possibly to Fc gamma I receptor.

Type: Primary
Antigen: Fcgr3; Fcgr2b
Clonality: Monoclonal
Clone: 2.4G2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge