Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD55 Chicken Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

XW-8099
PRSIXW-8099EA 826 EUR InStock
PRSIXW-8099
Anti-CD55 Chicken Polyclonal Antibody
Antikörper
FUNCTION: Also acts as the receptor for echovirus 7 and related viruses (echoviruses 13, 21, 29 and 33).
SUBUNIT: Monomer (major form) and non-disulfide-linked, covalent homodimer (minor form).
PTM: The Ser/Thr-rich domain is heavily O-glycosylated.
SIMILARITY: Contains 4 Sushi (CCP/SCR) domains.
SUBCELLULAR LOCATION: Attached to the membrane by a GPI-anchor (isoform 2).
TISSUE SPECIFICITY: Expressed on the plasma membranes of all cell types that are in intimate contact with plasma complement proteins. It is also found on the surfaces of epithelial cells lining extracellular compartments, and variants of the molecule are present in body fluids and in extracellular matrix.

Complement decay-accelerating factor antibody can be used for the detection of Complement decay-accelerating factor by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CD55
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge