Rough pump oil
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
22687
REST22687EA 48 EUR InStock
REST22687
Rough pump oil
Schmiermittel
This rough pump oil ideal for MSD and oil vacuum pumps is formulated from crude oil stocks known for their durability and line-lubricating qualities.
It can be used in Agilent 5973/5972/5971 and GCD mass spec systems or in other MSD systems that require rough pump oil.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge