30479
REST30479EA
63
EUR
InStock
REST30479
8260B matrix spike mix
Standards
Organische Standards
This mixture contains five components of 2500 µg/ml each in P and T methanol 1 ml/ampoule.
Components: Benzene, chlorobenzene, 1,1-dichloroethene, toluene and trichloroethylene.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.