31479
REST31479EA
49
EUR
InStock
REST31479
MA EPH surrogate spike mix (2 components)
Standards
Umweltanalyse-Standards
This spike mix contains two components: 1-Chlorooctadecane and o-Terphenyl.
Shelf life 6 months.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.