Method 525.2 PCB congener mix (8 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32420
REST32420EA 116.5 EUR InStock
REST32420
Method 525.2 PCB congener mix (8 components)
Standards Umweltanalyse-Standards
Contains eight components, those are 2-chlorobiphenyl, 2,3-dichlorobiphenyl, 2,4,5-trichlorobiphenyl, 2,2',4,4'-tetrachlorobiphenyl, 2,2',3',4,6-pentachlorobiphenyl, 2,2',4,4',5,6'-hexachlorobiphenyl, 2,2',3,3',4,4',6-heptachlorobiphenyl and 2,2',3,3',4,5',6,6'-octachlorobiphenyl.
Zertifizierungen: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge