32420
REST32420EA
116.5
EUR
InStock
REST32420
Method 525.2 PCB congener mix (8 components)
Standards
Umweltanalyse-Standards
Contains eight components, those are 2-chlorobiphenyl, 2,3-dichlorobiphenyl, 2,4,5-trichlorobiphenyl, 2,2',4,4'-tetrachlorobiphenyl, 2,2',3',4,6-pentachlorobiphenyl, 2,2',4,4',5,6'-hexachlorobiphenyl, 2,2',3,3',4,4',6-heptachlorobiphenyl and 2,2',3,3',4,5',6,6'-octachlorobiphenyl.
Zertifizierungen: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.