Accessories for ULT freezer TSX400V
Lieferant: HERAEUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FCR567TSX SS34567 FCR400TSX FCRP400TSX SK400TSX
HERAFCR567TSXEA 2020 EUR InStock
HERAFCR567TSX REVCSS34567 471-1171 471-1172 471-1173
Accessories for ULT freezer TSX400V
Gefriergeräte Ultratiefkühlgeräte
Record seven days of temperature performance at temperatures to −150 °C using Thermo Scientific™ chart recorders for ultra-low temperature freezers. Recorders use 150 mm chart paper and are available as factory or field installed.
  • Seven-day recorder
  • Field or factory installed
  • Field-installed units must be installed by a qualified professional
Order Now
PRODUKT
TECHNISCHE DATEN
SERVICELEISTUNGEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge