Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CHUK Mouse Monoclonal Antibody (Unconjugated) [clone: 14A231]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-301-H82
ROCK200-301-H82EA 1030 EUR InStock
ROCK200-301-H82
Anti-CHUK Mouse Monoclonal Antibody (Unconjugated) [clone: 14A231]
Antikörper
Anti-IKK alpha antibody is suitbale for use in Flow (I), IHC (p), IP and WB. Expect a band approximately 85-90kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1 µg/ml; Immunochemistry: 5 µg/ml; Immunoprecipitation: 1-2 µg/ml

Type: Primary
Antigen: CHUK
Clonality: Monoclonal
Clone: 14A231
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge