Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CCL20 Rat Monoclonal Antibody (Biotin) [clone: 4N5F7]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

210-506-B51
ROCK210-506-B51EA 856 EUR InStock
ROCK210-506-B51
Anti-CCL20 Rat Monoclonal Antibody (Biotin) [clone: 4N5F7]
Antikörper
This purified antibody has been tested for use in ELISA, IP, and Neutralization. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 0.5µg/ml;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: CCL20
Clonality: Monoclonal
Clone: 4N5F7
Conjugation: Biotin
Epitope: Full length
Host: Rat
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge