Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SNCA Mouse Monoclonal Antibody (Unconjugated) [clone: 3H9]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

500-301-E56
ROCK500-301-E56EA 976 EUR InStock
ROCK500-301-E56
Anti-SNCA Mouse Monoclonal Antibody (Unconjugated) [clone: 3H9]
Antikörper
Alpha Synuclein antibody is suitable for use in ELISA, Western Blotting and IF. Specific conditions should be optimized by the end user. Expect a band of approximately 15 kDa in size corresponding to the alpha synuclein Proteins in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000; Immunofluorescence: 1:500; Western Blot: 1:1000

Type: Primary
Antigen: SNCA
Clonality: Monoclonal
Clone: 3H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Rat, Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge