Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GSK3B Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

600-401-424
ROCK600-401-424EA 821 EUR InStock
ROCK600-401-424
Anti-GSK3B Rabbit Polyclonal Antibody
Antikörper
This antibody has been tested for use in ELISA, immunochemistry, western blotting and immunoprecipitation.  Reactivity in other immunoassays is unknown.  Serum starved 293T whole cell lysate is suitable for use as a positive control.  Anti-GSK 3B sho

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:200 - 1:1000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:200 - 1:1000;

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge