Tissue grinder, Potter-Elvehjem, ground glass, KIMBLE®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP885500-0019 SP885500-0021 SP885500-0022 SP885500-0023 SP885500-0024
SCERSP885500-0019EA 440 EUR InStock
SCERSP885500-0019 SCERSP885500-0021 SCERSP885500-0022 SCERSP885500-0023 SCERSP885500-0024
Tissue grinder, Potter-Elvehjem, ground glass, KIMBLE®
Homogenisatoren Gewebehomogenisatoren
Potter-Elvehjem with all-glass construction designed for a motor drive using a stirrer adapter.
  • Replacement components are available and interchangeable
  • Clearance between pestles and tubes is 0,004 to 0,006"
Zertifizierungen: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge