Ring light, VisiLED Slim
Lieferant: SCHOTT AG Lighting and imaging
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
400150
SCOC400150EA 826 EUR InStock
SCOC400150
Ring light, VisiLED Slim
Beleuchtung Ringlichter
The VisiLED Slim is a space saving LED ring light, optimal for unconfined object preparation or usage with objective revolvers.
  • Easily adaptable to microscope objectives by means of thumb screw
  • Homogeneous incident or oblique illumination
  • Controllable with all VisiLED controllers (MC750, MC1000, MC1500)
  • Variable working distance ranges possible by exchange of focus optic rings
  • Implementation of brightfield and darkfield applications
  • Segments controllable in different preset modes
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge