Drying time recorder for film applicators
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AB3500
SIMEAB3500EA 774.49 EUR InStock
SIMEAB3500
Drying time recorder for film applicators
Diagrammschreiber Diagrammschreiber
Drying time recorder tool can be used as an optional test tool for the automatic film applicators. This device allows the user to accurately define the drying time of various types of coatings within the time range of 1 minute to 48 hours. The drying time recorder option can be easily clamped on any standard automatic film applicator with firmware 2,01 and above.
  • With digital indication on drying stage
  • 'Intelligent recording' option significantly improves the resolution of longer drying process
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge