Water penetration tester, acc. to Karsten
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LI7500
SIMELI7500EA 408.24 EUR InStock
SIMELI7500
Water penetration tester, acc. to Karsten
Wasserpenetrations-Tester
This Karsten tube penetration test is a simple test for measuring the degree or water penetration into building materials such as concrete, stone and plaster. The test kit consists of a glass tube filled with water, bonded to the test material with plastiline. Water pressure is then exerted on the surface. A graduated scale indicates, over time, the amount or water penetrated into the surface.
  • Tube material: Glass
  • Suitable for vertical and horizontal surfaces
Bestellinformation: The TQC Karsten Tube Penetration Test contains 3 tubes for horizontal surfaces, 3 tubes for vertical surfaces (or at choice any other combination of total 6 tubes), a water bottle, marker and putty to place the tubes leak-tight on the surface. Additional vertical or horizontal tubes are available on request.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge