Reagent Reservoirs
Lieferant: Simport Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
R225-1 R225-15 R225-3 R225-5 R225-7 R225-9
SIMPR225-1EA 581 EUR InStock
SIMPR225-1 SIMPR225-15 SIMPR225-3 SIMPR225-5 SIMPR225-7 SIMPR225-9
Reagent Reservoirs
Reagenz-Reservoirs
For labs involved with fraction collecting, compound storage, plate replication, and mixing applications. Suitable for a range of liquid handling configurations from 8 or 12 channel pipettes to 96 or 384 pipette heads.
  • Robot friendly, designed for simple, economical handling of reagents in automated systems
  • Bottom designed to enable funneling of liquid, maximising reagent recovery with pipettes
  • Low-profile
  • Chemical and heat resistant, can be stored at temperatures as low as –80 °C
  • Autoclavable
  • Can be gamma irradiated
  • Certified DNase/RNase and pyrogen free
Ideal for small volume robotic tips used in high throughput instruments. Choose from a single reservoir or partitioned models for several liquids.
Conforms to ANSI/SLAS standard dimensions, they are compatible with virtually any automated liquid handler, stacker, and manual procedure.
Made from natural virgin polypropylene, providing a low binding surface to prevent samples from sticking to the sidewalls during elution and is chemically inert for combinatorial chemistry applications.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge