Drucken

Antibody labelling kits, HRP-Antibody All-in-One Conjugation kit

Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

A-9002-001
SOLUA-9002-001EA 1550 EUR
SOLUA-9002-001
Antibody labelling kits, HRP-Antibody All-in-One Conjugation kit
Tests Proteintests

The HRP-Antibody All-in-One Conjugation kit offers TurboLink™ catalysed conjugation that delivers 100% conversion of antibody to conjugate and purification that generates a conjugate free of unlabelled antibody or enzyme at a high final yield of >50%, without HPLC. The standard kit includes everything needed to make direct-labelled two HRP-antibody conjugates from 100 µg of any user-supplied antibody.


  • Higher signal – from better incorporation rate
  • Catalysed conjugation – fast catalysed method generates conjugates in just over 5 h
  • Quantifiable – conjugate bond is UV-traceable for simple and direct quantification
  • High purity – spin-column purification columns deliver 100% purified conjugates
  • Gentle – amine-reactive chemistry leaves antibodies intact - no reducing reagents
  • Easy to use – protocols and calculators provided to determine MSR
  • Versatile – compatible with Western, ELISA, or IHC applications
  • Efficient – 100% conversion with 50 to 70% yields

Order Now


Learn more