Disposable columns, RediSep® Rf
Lieferant: TELEDYNE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
69-2203-380
TELE69-2203-380EA 0 EUR InStock
TELE69-2203-380
Disposable columns, RediSep® Rf
Chromatographiesäulen
RediSep Rf disposable, bare silica columns for Flash purification of organic compounds are the benchmark for normal phase separations. This high quality media is specified for high resolution and reproducibility for everyday purifications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge