These safety storage cabinets for flammable liquids with a fire resistance up to 30 minutes are constructed with insulated panel in RAL 7035 (light grey) and doors either in RAL 7035 (light grey) or RAL 1023 (yellow plated). With standardised symbols.
- Melamine wood structure
- Has fire resistance up to 30 mins
- Height-adjustable shelves at intervals of 32 mm
- Base with cover for easy movement by truck (except for 'under-bench' models)
- Swing door with automatic closing
- Key locking in 3 points
- Adjustable feet for levelling
Suitable for all types of chemicals commonly used in laboratories and industries. Also used in the separation from families of hazardous products under the Code of Public Health and the recommendations of the I.N.R.S.
Models 3031 ARD and 3032 ARD feature a liquid waste recovery system.
Zertifizierungen: 3030 range type testing according to DIN EN 14470-1 and FM 6050. 7030 range testing according to DIN EN 14470-1.
Bestellinformation: Contact your Avantor representative for more information and options.