Safety storage cabinets for flammable liquids
Lieferant: TRIONYX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3032M11 3031E 3032E 3034E 3034M2 3034M3 3034M4 3035E 3035M11 3035M12 3035M13 3035M22
TRIO3032M11EA 2517 EUR InStock
TRIO3032M11 TRIO3031E TRIO3032E TRIO3034E TRIO3034M2 TRIO3034M3 TRIO3034M4 TRIO3035E TRIO3035M11 TRIO3035M12 TRIO3035M13 TRIO3035M22
Safety storage cabinets for flammable liquids
Möbel Lagerschränke
These safety storage cabinets for flammable liquids with a fire resistance up to 30 minutes are constructed with insulated panel in RAL 7035 (light grey) and doors either in RAL 7035 (light grey) or RAL 1023 (yellow plated). With standardised symbols.
  • Melamine wood structure
  • Has fire resistance up to 30 mins
  • Height-adjustable shelves at intervals of 32 mm
  • Base with cover for easy movement by truck (except for 'under-bench' models)
  • Swing door with automatic closing
  • Key locking in 3 points
  • Adjustable feet for levelling
Suitable for all types of chemicals commonly used in laboratories and industries. Also used in the separation from families of hazardous products under the Code of Public Health and the recommendations of the I.N.R.S.
Models 3031 ARD and 3032 ARD feature a liquid waste recovery system.
Zertifizierungen: 3030 range type testing according to DIN EN 14470-1 and FM 6050. 7030 range testing according to DIN EN 14470-1.
Bestellinformation: Contact your Avantor representative for more information and options.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge