Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Manual detergent, Emerald

Lieferant: TRISTEL SOLUTIONS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

CRY309 CRY310 CRY307
TRSTCRY309EA 1320 EUR InStock
TRSTCRY309 TRSTCRY310 141-0944
Manual detergent, Emerald
Reinigungsmittel Waschmittel und Oberflächenreiniger
A high foam, neutral, detergent for manual cleaning.

  • Neutral detergent for light to medium soiling
  • Dilution for use: Between 0,5% to 2% v/v

A manual detergent for the removal of light to medium soiling. Areas of use include walls, floors, surfaces, ceilings, equipment and utensils. Emerald is a soft metal safe detergent suitable for use on a variety of surfaces including plastics, silicone, vinyl and aluminium.

The crystel detergents are specifically formulated for the removal of the various residues that are present in the manufacturing and production process within pharmaceutical and personal care industries.

All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.

All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.

Lieferumfang: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.

Verpackung: Available in 5, 20, 200 and 1000 litre pack sizes.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge