Detergent, alkaline Cleaning-In-Place detergents, Amethyst and Topaz
Lieferant: TRISTEL SOLUTIONS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CRY402 CRY404 CRY405 CRY406 CRY403 CRY404/1L CRY401/1L
TRSTCRY402EA 0 EUR InStock
TRSTCRY402 TRSTCRY404 TRSTCRY405 TRSTCRY406 TRSTCRY403 TRSTCRY404/1L TRSTCRY401/1L
Detergent, alkaline Cleaning-In-Place detergents, Amethyst and Topaz
Reinigungsmittel Waschmittel und Oberflächenreiniger
A non foaming, medium duty caustic based, CIP detergent for the removal of organic soils, including fats and oils.
  • Medium duty detergent; for use on organic and inorganic soils
  • Suitable for use in CIP and spraywash enclosed systems
  • Dilution for use: amethyst - 1 to 5% v/v; topaz - 0,5 to 5% v/v
Areas of use include equipment such as CIP and spray wash enclosed systems. Amethyst and Topaz are not soft metal safe, but are suitable for use on stainless steel.
Crystel offer highly effective Cleaning-In-Place (CIP) detergents for automated processes within manufacturing sites. The product range is designed to remove a variety of residues that can be encountered. Residue test methods are available for all CIP detergents offering you peace of mind that there will be reduced risk of contamination of your products.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
Lieferumfang: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
Verpackung: Available in 20, 200 and 1000 litre pack sizes.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge