Disinfectants for instruments, Medistel
Lieferant: TRISTEL SOLUTIONS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MD307
TRSTMD307EA 0 EUR InStock
TRSTMD307
Disinfectants for instruments, Medistel
Reinigungsmittel Desinfektionsmittel
Medistel is used for chemical reprocessing of thermo-sensitive surgical instruments, flexible or rigid endoscopes and devices. It is suitable for both human and veterinary applications for disinfection of surgical instruments and medical devices. It is designed for open tank use and also suitable for other medical apparatus including ultrasonic baths.
  • Disinfects in 10 minutes and sterilises in 30 minutes
  • Dilution for use: 1:20
  • Compliant with BS EN ISO 9001:2000 and BS EN ISO 13485:2003
  • Conforms to medical directive 93/42/EEC requirements
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge