Combustion boats
Lieferant: TECHNICAL QUARTZ SOLUTIONS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
73375 73382 75140 75141 75142 75143 75144 75145 75146 75147
TSLA73375EA 82.1 EUR InStock
TSLA73375 TSLA73382 TSLA75140 TSLA75141 TSLA75142 TSLA75143 TSLA75144 TSLA75145 TSLA75146 TSLA75147
Combustion boats
Schiffchen Verbrennungsschiffchen
Combustion boats, quartz, glazed, with or without handle.
  • Very good chemical resistance
  • Very good temperature resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge