Quartz Glass Rods
Lieferant: TECHNICAL QUARTZ SOLUTIONS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
75350 75351 75352 75353 75354 75355
TSLA75350EA 22.9 EUR InStock
TSLA75350 TSLA75351 TSLA75352 TSLA75353 TSLA75354 TSLA75355
Quartz Glass Rods
Rührer Rührstäbe
Temperature resistant glass rods made from transparent quartz glass.
  • Chemical resistance
  • Made for tasks up to 1200 °C
  • High purity
TQS quartz glass rods stand for quality and versatile applications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge