BSA standard for protein assay
Lieferant: UPTIMA INTERCHIM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
UP36859A UP36859D
UPTIUP36859AEA 0 EUR InStock
UPTIUP36859A UPTIUP36859D
BSA standard for protein assay
Tests Proteintests
High quality, optimised reagents for the determination of protein concentration by the bicinchoninic acid method.
  • High sensitivity and wide linear dynamic range
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge