Sentry guard cartridges, XBridge™
Lieferant: WATERS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
186003064 186003061 186003060 186003079 186003082
WATE186003064EA 391 EUR InStock
WATE186003064 WATE186003061 WATE186003060 WATE186003079 WATE186003082
Sentry guard cartridges, XBridge™
Chromatographiesäulen
XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1 to 12). The trifunctionally bonded C18 ligand provides with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed. The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge