Reversed phase HPLC columns, Atlantis® T3
Lieferant: WATERS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
186003728 186003723 186003726 186003727 186003736 186003747 186003719 186003729
WATE186003728EA 1660 EUR InStock
WATE186003728 WATE186003723 WATE186003726 WATE186003727 WATE186003736 WATE186003747 WATE186003719 WATE186003729
Reversed phase HPLC columns, Atlantis® T3
Chromatographiesäulen
Atlantis T3 columns are universal, silica based, reversed phase C18 columns that provide balanced retention of polar and hydrophobic molecules and useful for developing separations of polar and non-polar compounds.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge