Solid phase extraction cartridges, Sep-Pak® Vac
Lieferant: WATERS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WAT043345 WAT036905 WAT020805 WAT054945 WAT043395 WAT054925 WAT054515 WAT020840 WAT020810
WATEWAT043345EA 511 EUR InStock
WATEWAT043345 WATEWAT036905 WATEWAT020805 WATEWAT054945 WATEWAT043395 WATEWAT054925 WATEWAT054515 WATEWAT020840 WATEWAT020810
Solid phase extraction cartridges, Sep-Pak® Vac
Chromatographiesäulen
These cartridges deliver the option of minimum sample preparation. Consists of moulded, high purity PP syringe-barrel bodies with two PE frits to contain the packing material.
  • Standard male Luer permits use with multi-position vacuum manifolds
  • Colour coded printing on syringe barrels for identification
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge