Wipes for general applications, Polytex® light
Lieferant: Zellstoff-Vertriebs-GmbH & Co. KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10160-00
ZVGA10160-00EA 144 EUR InStock
ZVGA10160-00
Wipes for general applications, Polytex® light
Wischtücher Reinigungstücher, wiederverwendbar Tücher für allgemeine Anwendungen
These solvent resistant wipes made of 100% PP are suitable for wet wiping in a variety of applications.
  • Durable and lint-free
  • Even remove stubborn dirt
  • Suitable for food contact
Ideal for use in garages, paint shops, catering areas, kitchens and food processing areas.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge