Food coats for men, 2206
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
2206WHRL 2206WHRM 2206WHRS 2206WHRXL 2206WHRXXL
PRTW2206WHRLEA 25 EUR InStock
PRTW2206WHRL PRTW2206WHRM PRTW2206WHRS PRTW2206WHRXL PRTW2206WHRXXL
Food coats for men, 2206
Arbeitskleidung
This food coat made from durable TradeGuard fabric (65% polyester, 35% cotton) is designed to withstand the busy tasks associated with food preparation environments.
  • Pre-shrunk, heavyweight fabric, hot head pressed
  • Inside chest pocket, two side pockets
  • Back buggy
  • Stud front, back vent
Length: 100 cm
Weight: 245 g/m²
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge