Capillary columns, TracePLOT™ TG-BOND S
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
26006-6000 26006-6030 26006-6090
555-8863EA 743 EUR InStock
555-8863 555-8862 555-8860
Capillary columns, TracePLOT™ TG-BOND S
Chromatographiesäulen
TracePLOT™ TG-BOND S GC columns are ideal for analysis of non-polar and mid-polar compounds.
  • Mid-polarity, divinylbenzene 4-vinylpyridine solid phase
  • Particles incorporated into the walls of the tubing for essentially no particle release
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge