HPLC columns, Hypersil™ SAX
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
34105-254630 34105-014001 34105-253030 34105-154630
555-5344EA 891 EUR InStock
555-5344 555-5362 555-5346 555-5348
HPLC columns, Hypersil™ SAX
Chromatographiesäulen
Hypersil™ SAX columns are ideal for the analysis of smaller organic molecules including nucleotides and organic acids.
  • High stability to aqueous and low pH mobile phases
  • Quaternary amine ion exchange ligand
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge