Sump trays, sheet steel
Lieferant: LACONT UMWELTTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
P25-1302-A P25-1303-A P25-1304-A P25-1305-A
LACOP25-1302-AEA 0 EUR InStock
LACOP25-1302-A LACOP25-1303-A LACOP25-1304-A LACOP25-1305-A
Sump trays, sheet steel
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Trays made of 3 mm galvanised sheet steel.
  • Suitable to place flat on the floor
Ideal for the storage of water-endangering and flammable liquids in small containers.
Bestellinformation: PE tray inserts are available should be order separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge