Syringe guides
Lieferant: HAMILTON BONADUZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
14806 14906
HAMI14806EA 57.1 EUR InStock
HAMI14806 HAMI14906
Syringe guides
Spritzen Spritzen für die Chromatographie
The syringe guide is a plunger support device. It is compatible with the 700, 1700 and 7000 series syringes. With only two guide arms the syringe guide protects the plunger from damage but should not be used to improve accuracy and precision.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge