ECD/FID make-up gas kit
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
21301 21302 27190 27191 27193 27192
REST21301EA 255 EUR InStock
REST21301 REST21302 REST27190 REST27191 REST27193 REST27192
ECD/FID make-up gas kit
Chromatographie-Zubehör Chromatographie-Ersatzteile
This kit is used exclusively with Agilent 5890 GCs.
Lieferumfang: The kit includes replacement fitting, ¹/₄" nut, Vespel®/graphite ferrule, ¹/₁₆" nut, 0,4 mm int. Ø graphite ferrule, Siltek®-treated guide.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge