Gas filters, Super Clean®
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
22019 21983 22021 22028 22029 22030 22031
REST22019EA 655 EUR InStock
REST22019 REST21983 REST22021 REST22028 REST22029 REST22030 REST22031
Gas filters, Super Clean®
Filter Gasfilter
Gas filter kits are designed for easy, leak tight cartridge changes. With glass inside to prevent diffusion and polycarbonate housing outside for safety.
Trap dimensions: 270×44 mm
Base plate dimensions: 102×102×48 mm
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge